Pyridine, BAKER, Laboratory Reagent, J.T.Baker®
Catalog # 8074.2500
Supplier: Avantor
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:2,5 L
- Seq:5096
Specifications
Specification Test Results
- Chloride (Cl):max. 0.005 %
- Residue after Evaporation:max. 0.005 %
- Water (H2O):max. 0.25 %
Specification Test Results
About this item
:
Merck Index:13,08060
Boiling Pt:115 °C (1013 hPa)
Melting Pt:–44 °C
Density:0.983 g/cm³ (20 °C)
Flash Pt:17 °C
Storage Temperature:Ambient
EINECS:203-809-9
UN:1282
ADR:3,II
:79.1 g/mol
:GHS02
:GHS07
CAS Number:110-86-1
Formula:C₅H₅N
MDL Number:MFCD00011732