

Pyridine, BAKER ANALYZED® ACS, J.T.Baker®
: Avantor
:110-86-1
:
:C₅H₅N
:MFCD00011732
:13,08060
:115 °C (1013 hPa)
:–44 °C
:0.983 g/cm³ (20 °C)
:17 °C
:1282
:3,II
:79.1 g/mol
:203-809-9
:GHS02
:GHS07
:Ambient
- Assay (by GC):min. 99.0 %
- Ammonia (NH3):max. 0.002 %
- Chloride (Cl):max. 0.001 %
- Reducing Substances:passes test
- Residue after Evaporation:max. 0.002 %
- Solubility in Water:passes test
- Suitability for OH groups determ. by phthalisation:passes test
- Sulfate (SO4):max. 0.001 %
- Water (H2O):max. 0.1 %
- Aluminium (Al):max. 0.5
- Boron (B):max. 0.1
- Barium (Ba):max. 0.1
- Calcium (Ca):max. 0.5
- Cadmium (Cd):max. 0.05
- Cobalt (Co):max. 0.02
- Chromium (Cr):max. 0.02
- Copper (Cu):max. 0.02
- Iron (Fe):max. 0.1
- Magnesium (Mg):max. 0.1
- Manganese (Mn):max. 0.02
- Nickel (Ni):max. 0.02
- Lead (Pb):max. 0.1
- Tin (Sn):max. 0.1
- Zinc (Zn):max. 1.0
Frequently Bought Together