

Pyridin 99+% ACS
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:110-86-1
:
Formula:C₅H₅N
MDL Number:MFCD00011732
Merck Index:13,08060
Boiling Pt:115…116 °C (760 mmHg)
Melting Pt:–42 °C
Density:0,978 g/cm³
Flash Pt:17 °C
UN:1282
ADR:3,II
:79.1 g/mol
EINECS:203-809-9
:GHS07
:GHS02
Storage Temperature:Stue temperatur
Specification Test Results
- Appearance:Clear colorless Liquid
- Water:≤0.1 % (K.F.)
- Solubility:(in water) Passes test
- GC:≥99.0 %
- Residue after evaporation:≤0.002 %
- Chloride (Cl):≤10 ppm
- Sulfate (SO4):≤10 ppm
- Copper (Cu):≤5 ppm
- Reducing substances:Passes test
- Ammonia (NH3):≤20 ppm
Specification Test Results
Frequently Bought Together