Overhead stirrer shafts, with blade, BOLA
Supplier: Bohlender
Certificates
About this item
Stirrer shafts made of PTFE-jacketed stainless steel with the blade made completely of PTFE.
- Chemical resistant
- Temperature resistance from –200 to +250 °C
- Creates tangential flow with little turbulence
Frequently Bought Together