
3,5-Dichloropyridine ≥98.0% (by GC)
Catalog # TCIAD0411-25G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:65.0 to 67.0 deg-C
Specification Test Results
About this item
:
:147.99 g/mol
Boiling Pt:178°C
Melting Pt:64…65 °C
EINECS:219-537-9
ADR:6.1,III
:GHS07
CAS Number:2457-47-8
Formula:C₅H₃Cl₂N
MDL Number:MFCD00006376