Order Entry
Denmark
Orders LinkContactUsLinkComponent
Glove liners, Perfect Poly® PU First White
Supplier:   HONEYWELL SAFETY
Glove liners, Perfect Poly® PU First White
Supplier:  HONEYWELL SAFETY
Certificates
Glove liners, Perfect Poly® PU First White
6 Options Available Below
Product Details & Documents
100% Nylon knitted glove with white PU coating on the palm and fingers.
  • Ventilated back optimises the glove's breathability
  • PU coating ensures a good grip at the fingertips
  • White colour enhances the glove's hygiene and cleanliness
Ideal for fine handling or as an underglove.
 : EN 388:2016 (3131X), Cat. II.
Specifications for Product Family
Specifications
  • Catalog No:
  • DALL2132255/6
  • DALL2132255/7
  • DALL2132255/8
  • DALL2132255/9
  • DALL2132255/10
  • DALL2132255/11
  • Ambidextrous / hand-specific:
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Disposable / reusable:
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Liner material:
  • Nylon
  • Nylon
  • Nylon
  • Nylon
  • Nylon
  • Nylon
  • Finish:
  • Coating on palm and fingers
  • Coating on palm and fingers
  • Coating on palm and fingers
  • Coating on palm and fingers
  • Coating on palm and fingers
  • Coating on palm and fingers
  • Gauge:
  • 13
  • 13
  • 13
  • 13
  • 13
  • 13
  • Cuff style:
  • Knit wrist
  • Knit wrist
  • Knit wrist
  • Knit wrist
  • Knit wrist
  • Knit wrist
  • EN norm:
  • EN 388:2016 (3131X)
  • EN 388:2016 (3131X)
  • EN 388:2016 (3131X)
  • EN 388:2016 (3131X)
  • EN 388:2016 (3131X)
  • EN 388:2016 (3131X)
Specifications
Product Family Options
Product Information
  • Glove size
    Colour
    Length
    Availability
    Price
  • 6
    White
    240 mm
    Log in for availability
    Log in for availability
    1 * 10 Par
    1 * 10 Par - 186,00 kr
    186,00 kr
    1 * 10 Par
       186,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    DALL2132255/6
    Ambidextrous / hand-specific
    Hand-specific
    Disposable / reusable
    Reusable
    Liner material
    Nylon
    Finish
    Coating on palm and fingers
    Gauge
    13
    Cuff style
    Knit wrist
    EN norm
    EN 388:2016 (3131X)
  • 7
    White
    240 mm
    Log in for availability
    Log in for availability
    1 * 10 Par
    1 * 10 Par - 186,00 kr
    186,00 kr
    1 * 10 Par
       186,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    DALL2132255/7
    Ambidextrous / hand-specific
    Hand-specific
    Disposable / reusable
    Reusable
    Liner material
    Nylon
    Finish
    Coating on palm and fingers
    Gauge
    13
    Cuff style
    Knit wrist
    EN norm
    EN 388:2016 (3131X)
  • 8
    White
    240 mm
    Log in for availability
    Log in for availability
    1 * 10 Par
    1 * 10 Par - 186,00 kr
    186,00 kr
    1 * 10 Par
       186,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    DALL2132255/8
    Ambidextrous / hand-specific
    Hand-specific
    Disposable / reusable
    Reusable
    Liner material
    Nylon
    Finish
    Coating on palm and fingers
    Gauge
    13
    Cuff style
    Knit wrist
    EN norm
    EN 388:2016 (3131X)
  • 9
    White
    240 mm
    Log in for availability
    Log in for availability
    1 * 10 Par
    1 * 10 Par - 186,00 kr
    186,00 kr
    1 * 10 Par
       186,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    DALL2132255/9
    Ambidextrous / hand-specific
    Hand-specific
    Disposable / reusable
    Reusable
    Liner material
    Nylon
    Finish
    Coating on palm and fingers
    Gauge
    13
    Cuff style
    Knit wrist
    EN norm
    EN 388:2016 (3131X)
  • 10
    White
    240 mm
    Log in for availability
    Log in for availability
    1 * 10 Par
    1 * 10 Par - 186,00 kr
    186,00 kr
    1 * 10 Par
       186,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    DALL2132255/10
    Ambidextrous / hand-specific
    Hand-specific
    Disposable / reusable
    Reusable
    Liner material
    Nylon
    Finish
    Coating on palm and fingers
    Gauge
    13
    Cuff style
    Knit wrist
    EN norm
    EN 388:2016 (3131X)
  • 11
    White
    240 mm
    Log in for availability
    Log in for availability
    1 * 10 Par
    1 * 10 Par - 186,00 kr
    186,00 kr
    1 * 10 Par
       186,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    DALL2132255/11
    Ambidextrous / hand-specific
    Hand-specific
    Disposable / reusable
    Reusable
    Liner material
    Nylon
    Finish
    Coating on palm and fingers
    Gauge
    13
    Cuff style
    Knit wrist
    EN norm
    EN 388:2016 (3131X)