Order Entry
Denmark
ContactUsLinkComponent
Naphthalene
Health
Irritant
Environment
Naphthalene
Catalog # PROOCHE158
CAS Number:  
undefined
Health
Irritant
Environment
Naphthalene
Catalog # PROOCHE158
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :128.17 g/mol
Merck Index:12,06457
Boiling Pt:218 °C (1013 hPa)
Melting Pt:79…82 °C
Density:1.145 g/cm³ (20 °C)
Flash Pt:79 °C
EINECS:202-049-5
UN:1334
ADR:4.1,III
 :GHS08
 :GHS07
 :GHS09
Storage Temperature:Ambient
CAS Number:91-20-3
Formula:C₁₀H₈
MDL Number:MFCD00001742