Order Entry
Denmark
ContactUsLinkComponent
Automated cell counter, Moxi™ Z
Automated cell counter, Moxi™ Z
Catalog # 734-2477
Supplier:  Orflo
undefined
Automated cell counter, Moxi™ Z
Catalog # 734-2477
Supplier:  Orflo

Specifications

  • Power:
    Battery (Lithium Ion 4400 mAc) or AC power (100 - 240 V, 50/60 Hz, 0,2 Amps)
  • Resolution:
    1200 histogram bins
  • Description:
    Cell counter
  • Data storage:
    500
  • Cell size:
    2 − 34 µm
  • W×D×H:
    190×108×70 mm
  • Output:
    Bluetooth and USB
  • Software:
    MoxiChart software package for Mac/PC
  • Pk:
    1 KIT

Specifications

About this item

Moxi Z is a mini automated cell counter that combines the Coulter principle used in high-end instruments with a patented thin-film sensor technology to provide highly accurate and repeatable particle sizing and counting over a broad dynamic range.

  • Particulate measurement in mammalian cells, RBC, WBC, common yeast
  • Measurement time of eight or 15 seconds
  • Very small: Fits in the palm of the hand
  • Long-life battery and LCD Touch
  • Bluetooth connectivity to MAC/PC for further analysis with MoxiChart Software

It also provides a reagent-free assessment of mammalian cell culture viability using a sophisticated electronic algorithm to report the standardized Moxi Viability Index (MVI).

NOTE: Users can choose from type M (suitable for cell sizes in the range of 4 to 26 µm) or type S (suitable for cell sizes in the range 3 to 20 µm) cassettes depending on the cell specifications.

Please note: Users can choose from Type M (suitable for cell sizes in the range of 4-26 µm) or Type S (suitable for cell sizes in the range 3-20 µm) cassettes depending on the cell specifications.

Certifications: TÜV certified
Delivery: Supplied with 25 cassettes (sufficient for 50 tests), cassette dispenser, calibration check beads, USB cable/power adapter and USB flash drive containing user guide and MoxiChart software.