Order Entry
Denmark
Orders LinkContactUsLinkComponent
Silver nitrate

Silver nitrate

Oxidizing
Corrosive
Environment
Marketsource
Supplier:   Serva
Silver nitrate
Supplier:  Serva
Marketsource
Certificates
Silver nitrate

3 Options Available Below

Product Details & Documents
Synonyms:  Lunar caustic, Silver mononitrate, Nitric acid silver salt
  • CAS Number:7761-88-8
  • Formula:AgNO₃
  • MDL Number:MFCD00003414
  • Molecular Weight:169.87 g/mol
  • Merck Index:12,08661
  • Boiling Pt:444 °C (1013 hPa)
  • Melting Pt:210 °C
  • Density:4.35 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:231-853-9
  • UN:1493
  • ADR:5.1,II
  • REACH:01-2119513705-43
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • 25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 537,67 kr
    537,67 kr
    1 * 25 g
       537,67 kr

    Specifications:
    Marketsource
    Marketsource
  • 100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 1.650,35 kr
    1.650,35 kr
    1 * 100 g
       1.650,35 kr

    Specifications:
    Marketsource
    Marketsource
  • Plastic bottle for solids
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g

    Specifications:
    Marketsource
    Marketsource
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.