Order Entry
Denmark
Orders LinkContactUsLinkComponent
Silver nitrate
Marketsource
 :   Roth Carl
Silver nitrate
 :  Roth Carl
Marketsource
Silver nitrate
Silver nitrate
 :  Lunar caustic, Silver mononitrate, Nitric acid silver salt
  •  :7761-88-8
  •  :AgNO₃
  •  :MFCD00003414
  •  :169.87 g/mol
  •  :12,08661
  •  :444 °C (1013 hPa)
  •  :210 °C
  •  :4.35 g/cm³ (20 °C)
  •  :Ambient
  •  :231-853-9
  •  :1493
  •  :5.1,II
  •  :01-2119513705-43
Product Family Options
  • Pk
  • 50 g
    1 * 50 g
    1 * 50 g
    Marketsource
    Marketsource
  • 250 g
    1 * 250 g
    1 * 250 g
    Marketsource
    Marketsource
  • 1 kg
    1 * 1 kg
    1 * 1 kg
    Marketsource
    Marketsource