Phosphorus(V) chloride 98%
:10026-13-8
:
:Cl₅P
:MFCD00011439
:208.24 g/mol
:12,07508
:167 °C (subl.)
:1.6 g/cm³
:233-060-3
:1806
:8,II
:GHS06
:GHS08
:GHS05
:Ambient
- Appearance:Yellow Powder and/or scales
- Titration Argentometric:≥97.5 %
- Melting point:38 to 44 °C
- GC:≥96.0 %
Frequently Bought Together