Order Entry
Denmark
ContactUsLinkComponent
Silica gel, beads ~2 - 5 mm desiccant, Supelco®
Silica gel, beads ~2 - 5 mm desiccant, Supelco®
Catalog # 1.07735.1000
Supplier:  Merck
CAS Number:  
Silica gel, beads ~2 - 5 mm desiccant, Supelco®
Catalog # 1.07735.1000
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 kg
  • Seq:
    4024
  • Environmentally Preferable:

Specifications

About this item

 :
 :60.08 g/mol
Boiling Pt:2230
Melting Pt:1710
Storage Temperature:Ambient
EINECS:231-545-4
REACH:01-2119379499-16
CAS Number:63231-67-4
 :112926-00-8
Formula:SiO₂
MDL Number:MFCD00011232
 :M-Clarity™ quality level = MQ200