

Orthoboric acid 99.5-100.5% (dry basis), granular NF, J.T.Baker®
Supplier: Avantor
Certificates
About this item
CAS Number:10043-35-3
:
Formula:B(OH)₃
MDL Number:MFCD00011337
Merck Index:13,01326
Boiling Pt:185 °C (1013 hPa)
Melting Pt:160 °C
Density:1.44 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:233-139-2
ADR:5.1,III
:61.83 g/mol
REACH:01-2119486683-25
:GHS07
:GHS08
Specification Test Results
- GMP Manufactured Product:Passes Test
- Meets N.F. Requirements:Passes Test
- CAUTION: For Manufacturing, processing or repackaging:≤0.5%
- Bulk Pharmaceutical Chemical:99.5 - 100.5%
- NF - Completeness of Solution:Passes Test
- NF - Identification:Passes Test
- NF - Solubility in Alcohol:Passes Test
- NF - Loss on Drying:≤0.5 %
- NF - Assay (H₃BO₃) (dried basis):99.5 - 100.5 %
Specification Test Results
Frequently Bought Together