Order Entry
Denmark
ContactUsLinkComponent
 
3-Methyl-1-phenyl-2-pyrazolin-5-one, Millipore®
Catalog # 443300-500
CAS Number:  
3-Methyl-1-phenyl-2-pyrazolin-5-one, Millipore®
Catalog # 443300-500
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 mg
  • Seq:
    2725

Specifications

About this item

 :
 :174.2 g/mol
EINECS:201-891-0
CAS Number:89-25-8
Formula:C₁₀H₁₀N₂O
MDL Number:MFCD00003138