1,4-Butanediol diglycidyl ether 96%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:2425-79-8
:
Formula:C₁₀H₁₈O₄
MDL Number:MFCD00005146
:202.25 g/mol
Boiling Pt:157…158 °C (11 mmHg)
Density:1.049
Flash Pt:140 °C (284 °F)
EINECS:219-371-7
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together