1,4-Bis(hydroxymethyl)cyclohexane (mixture of cis and trans isomers) ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: 1,4-Cyclohexanedimethanol
CAS Number:105-08-8
:
Formula:C₈H₁₆O₂
MDL Number:MFCD00001512
Boiling Pt:282…284 °C
Melting Pt:∼ 30 °C
Density:1.04
Molecular Weight:144.21 g/mol
Flash Pt:161 °C (322 °F)
Storage Temperature:Ambient
EINECS:203-268-9
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together