Tetraethylene glycol dimethyl ether ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: 2,5,8,11,14-Pentaoxapentadecane
CAS Number:143-24-8
:
Formula:C₁₀H₂₂O₅
MDL Number:MFCD00008505
:222.28 g/mol
Merck Index:14,9208
Boiling Pt:275…276 °C
Melting Pt:–30 °C
Density:1.01
Flash Pt:140 °C (284 °F)
EINECS:205-594-7
:GHS08
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together