Order Entry
Denmark
ContactUsLinkComponent
Palladium 10% on activated carbon, dry, unreduced
Flammable
Palladium 10% on activated carbon, dry, unreduced
Certificates
undefined
Flammable
Palladium 10% on activated carbon, dry, unreduced

Specification Test Results

  • Appearance:
    Black Powder
  • Assay:
    9.7 to 10.3 % (Pd)
  • Loss on drying:
    ≤5 %

Specification Test Results

Related Information

Frequently Bought Together