Anthrone ACS
Leverandør: Thermo Fisher Scientific
Om denne vare
Synonymer: Anthracen-9(10H)-one
- CAS nummer:90-44-8
- Formel:C₁₄H₁₀O
- MDL nummer:MFCD00001187
- Molecular Weight:194.23 g/mol
- Kogende Pt:721 °C (760 mmHg)
- Smeltende Pt:152…157 °C
- Opbevaringstemperatur:Ambient
- EINECS:201-994-0
2 tilgængelige muligheder nedenfor
Product Details & Documents
Synonymer: Anthracen-9(10H)-one
- CAS nummer:90-44-8
- Formel:C₁₄H₁₀O
- MDL nummer:MFCD00001187
- Molecular Weight:194.23 g/mol
- Kogende Pt:721 °C (760 mmHg)
- Smeltende Pt:152…157 °C
- Opbevaringstemperatur:Ambient
- EINECS:201-994-0
Documents
Specifications for Product Family
Specifikationstestresultater
- Appearance:Yellow Crystals
- Infrared spectrum:Conforms
- Melting point:152 to 157 °C
- Solubility:(in ethyl acetate) Passes test
- Absorbance:≤0.75 (425 nm, 1 cm cell)
- Sensitivity:(to carbohydrates) Passes test
Product Family Options
Produktinformation
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



