Order Entry
Denmark
ContactUsLinkComponent
Barium chloride, Suprapur® for trace analysis, Supelco®
Barium chloride, Suprapur® for trace analysis, Supelco®
Supplier:  Merck
Certificates
Barium chloride, Suprapur® for trace analysis, Supelco®
Supplier:  Merck
Certificates

Specification Test Results

  • Solubility:
    375 g/l (20 °C)
  • Melting Point:
    963 °C
  • Molar Mass:
    208.23 g/mol
  • Bulk Density:
    1350 kg/m3
  • Boiling Point:
    1560 °C
  • Density:
    3.9 g/cm3 (20 °C)
  • pH:
    5 - 8 (50 g/l, H2O, 20 °C)
  • Hazard Class:
    6.1
  • Hazard Class Description:
    Toxic Materials

Specification Test Results

Related Information

Frequently Bought Together