Order Entry
Denmark
ContactUsLinkComponent
Sodium selenite Se 44-46%, anhydrous
Sodium selenite Se 44-46%, anhydrous
Certificates
undefined
Sodium selenite Se 44-46%, anhydrous
Certificates

Specification Test Results

  • Appearance:
    White to light beige Powder
  • Assay:
    44 to 46 % (Se)
  • Loss on drying:
    ≤0.5 %
  • Iron (Fe):
    ≤300 ppm
  • Lead (Pb):
    ≤500 ppm

Specification Test Results

Related Information

Certificates
Search by Lot Number
Frequently Bought Together