Order Entry
Denmark
Orders LinkContactUsLinkComponent
Centrifuge tubes
Centrifuge tubes
Supplier:   MARIENFELD
Centrifuge tubes
Supplier:  MARIENFELD
Certifikater
Centrifuge tubes
Centrifuge tubes
2 tilgængelige muligheder nedenfor
Product Details & Documents
Borosilicate glass 3,3.
  • Heavy walled
  • Ungraduated
  • Very good chemical resistance
  • Very good temperature resistance
Short conical or round bottom.
Product Family Options
Produktinformation
  • Capacity
    Ø×L
    Closure type
    Type
    Availability
    Price
  • 10 ml
    16/17×98 mm
    Without cap
    Conical bottom
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 100 Items
    1 * 100 Items - 797,00 kr
    797,00 kr
    1 * 100 Items
       797,00 kr
  • 50 ml
    34×100 mm
    Without cap
    Round bottom
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 10 Items
    1 * 10 Items - 605,00 kr
    605,00 kr
    1 * 10 Items
       605,00 kr