Sodium metabisulphite
Marketsource
Supplier: Roth Carl
Jump to:
About this item
Synonyms: Disodium disulphite, di-Sodium disulphite, Sodium pyrosulphite, Disodiumdisulphite, Sodium disulphite
- CAS Number:7681-57-4
- Formula:Na₂S₂O₅
- MDL Number:MFCD00167602
- Molecular Weight:190.11 g/mol
- Merck Index:12,08784
- Melting Pt:150 °C
- Density:1.48 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:231-673-0
- REACH:01-2119531326-45
3 Options Available Below
Product Details & Documents
Synonyms: Disodium disulphite, di-Sodium disulphite, Sodium pyrosulphite, Disodiumdisulphite, Sodium disulphite
- CAS Number:7681-57-4
- Formula:Na₂S₂O₅
- MDL Number:MFCD00167602
- Molecular Weight:190.11 g/mol
- Merck Index:12,08784
- Melting Pt:150 °C
- Density:1.48 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:231-673-0
- REACH:01-2119531326-45
Product Family Options
Product Information
- Pack typePkAvailabilityPrice
- Specifications:MarketsourceMarketsource
- Specifications:MarketsourceMarketsource
- Specifications:MarketsourceMarketsource
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



