Order Entry
Denmark
Orders LinkContactUsLinkComponent
Sodium thiosulphate

Sodium thiosulphate

50
Catalog # HACH2293766
Supplier:   Hach
Sodium thiosulphate
Catalog # HACH2293766
Supplier:  Hach
CAS Number:  7772-98-7
Sodium thiosulphate

50

CAS Number:  7772-98-7
Price
253,00 kr
Log in for availability
1 * 50 Items
Product Details & Documents
Synonyms:  Sodiumthiosulphate, Sodium hyposulphite, Antichlor, di-Sodium thiosulphate
  • CAS Number:7772-98-7
  • Formula:Na₂S₂O₃
  • MDL Number:MFCD00003499
  • Molecular Weight:158.11 g/mol
  • Merck Index:12,08844
  • Boiling Pt:100 °C (1013 hPa)
  • Melting Pt:48 °C
  • Density:1.66 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:231-867-5
Specifications

Specifications

  • Pk:
    50

Specifications