Order Entry
Denmark
ContactUsLinkComponent
Sodium hydroxide ≥97.0% (by titrimetric analysis), pellets, Purum p.a.
Corrosive
Sodium hydroxide ≥97.0% (by titrimetric analysis), pellets, Purum p.a.
Catalog # 71692-5KG
CAS Number:  
Corrosive
Sodium hydroxide ≥97.0% (by titrimetric analysis), pellets, Purum p.a.
Catalog # 71692-5KG
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    5 kg
  • Seq:
    4114

Specifications

Specification Test Results

  • Vapor Pressure:
    <18 mmHg ( 20 °C)
  • Assay:
    Min. 97 %
  • Assay of Na₂CO₃:
    Max. 0.8 %
  • Silver (Ag):
    Max. 0.001 %
  • Aluminium (Al):
    Max. 0.001 %
  • Calcium (Ca):
    Max. 0.005 %
  • Cadmium (Cd):
    Max. 0.005 %
  • Cobalt (Co):
    Max. 0.005 %
  • Copper (Cu):
    Max. 0.005 %
  • Iron (Fe):
    Max. 0.001 %
  • Mercury (Hg):
    Max. 0.00001 %
  • Potassium (K):
    Max. 0.1 %
  • Nickel (Ni):
    Max. 0.001 %
  • Lead (Pb):
    Max. 0.001 %
  • Zinc (Zn):
    Max. 0.005 %
  • Chloride (Cl):
    Max. 0.005 %
  • Total N:
    Max. 0.0005 %
  • Phosphate (PO₄):
    Max. 0.001 %
  • Silicate (as SiO₂):
    Max. 0.001 %
  • Sulfate (SO₄):
    Max. 0.005 %

Specification Test Results

About this item

 :
 :40 g/mol
Merck Index:14,08627
Boiling Pt:1.390 °C (1.013 hPa)
Melting Pt:319 °C
Density:2.13 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:215-185-5
UN:1823
ADR:8,II
REACH:01-2119457892-27
 :GHS05
CAS Number:1310-73-2
Formula:NaOH
MDL Number:MFCD00003548