Order Entry
Denmark
Orders LinkContactUsLinkComponent
Sodium hydroxide for elementary analysis

Sodium hydroxide for elementary analysis

Glass bottle for solids, 500 g
Catalog # ELTA90210
Corrosive
Supplier:   ELTRA
Sodium hydroxide for elementary analysis
Catalog # ELTA90210
Supplier:  ELTRA
CAS Number:  1310-73-2
Sodium hydroxide for elementary analysis

Glass bottle for solids, 500 g

CAS Number:  1310-73-2
Price
1.550,00 kr
Log in for availability
1 * 500 g
Product Details & Documents
Synonyms:  Caustic soda
  • CAS Number:1310-73-2
  • Formula:NaOH
  • MDL Number:MFCD00003548
  • Molecular Weight:40 g/mol
  • Merck Index:14,08627
  • Boiling Pt:1390 °C (1013 hPa)
  • Melting Pt:323 °C
  • Density:2.13 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:215-185-5
  • UN:1823
  • ADR:8,II
  • REACH:01-2119457892-27

Documents

Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g

Specifications