Sodium hydroxide, BAKER, J.T.Baker®
Supplier: Avantor
About this item
Synonyms: Caustic soda
- CAS Number:1310-73-2
- Formula:NaOH
- MDL Number:MFCD00003548
- Molecular Weight:40 g/mol
- Merck Index:14,08627
- Boiling Pt:1390 °C (1013 hPa)
- Melting Pt:323 °C
- Density:2.13 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:215-185-5
- UN:1823
- ADR:8,II
- REACH:01-2119457892-27
3 Options Available Below
Product Details & Documents
Synonyms: Caustic soda
- CAS Number:1310-73-2
- Formula:NaOH
- MDL Number:MFCD00003548
- Molecular Weight:40 g/mol
- Merck Index:14,08627
- Boiling Pt:1390 °C (1013 hPa)
- Melting Pt:323 °C
- Density:2.13 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:215-185-5
- UN:1823
- ADR:8,II
- REACH:01-2119457892-27
Documents
Certificates
Search by Lot Numberspec_BAKR0405.1000.pdf spec_BAKR0405.5000.pdf spec_BAKR0405.9025.pdf
Product specification
Specifications for Product Family
Specification Test Results
- Assay:97.0-100.5 %
- Appearance of solution:passes test
- Carbonate (as Na2CO3):max. 2,0 %
- Chlorides (as Cl):max. 50 ppm
- Heavy Metals (as Pb):max. 20 ppm
- Identification:passes test
- Iron (Fe):max. 10 ppm
- Sulfates (as SO4):max. 50 ppm
- Additional Specification(s)::-
- Potassium (K):passes test



