Ground joint tube fittings, for pressure upto 5 Bar, BOLA
PTFE, white. For connecting tubes or tubing to vessels with ground joint. For inserting and fixing probes, thermometers, dip tubes or cables.
- Temperature resistance: −200 to +250 °C
- Universal chemical resistance
- Autoclavable
- Vacuum: suitable
Fitting made of PTFE for transition from ground joints to metric threads for connecting hard-walled tubing (e.g. PTFE, PFA or FEP). With nuts made of glass-fibre reinforced PTFE, body with rings and knurled grip for opening. The product is only exposed to PTFE.
Frequently Bought Together