Order Entry
Denmark
ContactUsLinkComponent
Sodium amide
Flammable
Corrosive
Sodium amide
Catalog # MOLEM58630340
Supplier:  Molekula
CAS Number:  
Flammable
Corrosive
Sodium amide
Catalog # MOLEM58630340
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g
  • Seq:
    3645

Specifications

About this item

 :
 :39.01 g/mol
Merck Index:12,08717
Boiling Pt:400 °C (1013 hPa)
Melting Pt:208 °C
Density:1.39 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:231-971-0
UN:1390
ADR:4.3,II
 :GHS02
 :GHS05
CAS Number:7782-92-5
Formula:H₂NNa
MDL Number:MFCD00011117