Sodium azide
Marketsource
Supplier: Serva
Jump to:
About this item
Synonyms: Sodium triazide
- CAS Number:26628-22-8
- Formula:NaN₃
- MDL Number:MFCD00003536
- Molecular Weight:65.01 g/mol
- Merck Index:13,08653
- Melting Pt:275 °C
- Density:1.85 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:247-852-1
- UN:1687
- ADR:6.1,II
3 Options Available Below
Product Details & Documents
Synonyms: Sodium triazide
- CAS Number:26628-22-8
- Formula:NaN₃
- MDL Number:MFCD00003536
- Molecular Weight:65.01 g/mol
- Merck Index:13,08653
- Melting Pt:275 °C
- Density:1.85 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:247-852-1
- UN:1687
- ADR:6.1,II
Product Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



