Potassium permanganate
Catalog # MOLEM33605494
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
- Environmentally Preferable:
Specifications
About this item
Synonyms: Permanganic acid potassium salt
:
:158.03 g/mol
EINECS:231-760-3
UN:1490
:GHS03
:GHS07
:GHS09
Storage Temperature:Ambient
CAS Number:7722-64-7
Formula:KMnO₄
MDL Number:MFCD00011364
Melting Pt:> 240 °C
Density:2.7 g/cm³ (20 °C)
ADR:5.1 II
Merck Index:12,07824