Order Entry
Denmark
ContactUsLinkComponent
Potassium permanganate 99.0-100.5% (by manganometry), EMSURE® Reag. Ph. Eur., ACS for analysis, Supelco®
Potassium permanganate 99.0-100.5% (by manganometry), EMSURE® Reag. Ph. Eur., ACS for analysis, Supelco®
Supplier:  Merck
Certificates
Potassium permanganate 99.0-100.5% (by manganometry), EMSURE® Reag. Ph. Eur., ACS for analysis, Supelco®
Supplier:  Merck
Certificates

Specifications

  • Environmentally Preferable:

Specifications

Specification Test Results

  • Assay (manganometric):
    99.0 - 100.5 %
  • Identity:
    Passes test
  • Appearance of solution:
    Passes test
  • In water insoluble matter:
    ≤ 0.2 %
  • Chloride (Cl):
    ≤ 0.02 %
  • Chloride, Chlorate (as Cl):
    ≤ 0.005 %
  • Sulfate (SO₄):
    ≤ 0.01 %
  • Total nitrogen (N):
    ≤ 0.003 %
  • Cu (Copper):
    ≤ 0.001 %
  • Fe (Iron):
    ≤ 0.002 %
  • Pb (Lead):
    ≤ 0.002 %

Specification Test Results

Related Information

Frequently Bought Together