Potassium iodate 99.4-100.4% ACS
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7758-05-6
:
Formula:KlO₃
MDL Number:MFCD00011406
Density:3.89
Molecular Weight:214 g/mol
Melting Pt:560 °C (dec.)
ADR:5.1 II
Merck Index:14,7642
EINECS:231-831-9
UN:1479
:GHS03
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together