
Lithium fluoride, Suprapur® for trace analysis, Supelco®
Catalog # 1.05686.0050
Supplier: Merck
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:50 g
- Seq:3368
Specifications
Specification Test Results
- Solubility:1.3 g/l (25 °C)
- Melting Point:870 °C
- Molar Mass:25.94 g/mol
- Bulk Density:600 kg/m3
- Boiling Point:1676 °C (1013 hPa)
- Density:2.64 g/cm3 (20 °C)
- Hazard Class:6.1
- Hazard Class Description:Toxic Materials
Specification Test Results
About this item
:
:25.94 g/mol
Merck Index:13,05553
Boiling Pt:1676 °C (1013 hPa)
Melting Pt:870 °C
Density:2.64 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:232-152-0
UN:3288
ADR:6.1,III
:M-Clarity™ quality level = MQ100
:GHS07
CAS Number:7789-24-4
Formula:FLi
MDL Number:MFCD00011090