Potassium chromate ≥99.5%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:7789-00-6
:
Formula:K₂CrO₄
MDL Number:MFCD00011368
:194.19 g/mol
Merck Index:12,07784
Boiling Pt:1000 °C (1013 hPa)
Melting Pt:985 °C
Density:2.73 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:232-140-5
UN:3087
ADR:5.1 (6.1),III
:GHS08
:GHS07
:GHS09
Specification Test Results
- Assay:Min. 99.5 %
- pH (20°C; 5 %):8.6 - 9.8
- Insolubility in water:Max. 50 ppm
- Cl (Chloride):Max. 10 ppm
- SO4 (Sulphate):Max. 100 ppm
- Ca (Calcium):Max. 50 ppm
- Cu (Copper):Max. 10 ppm
- Fe (Iron):Max. 15 ppm
- Na (Sodium):Max. 0.03 %
- Pb (Lead):Max. 50 ppm
Specification Test Results
Related Information
Certificates
Search by Lot Number250 ml Plastic bottle for solids DIN 60 (ZPACK13250250) 750 ml Plastic bottle for solids DIN 60 (ZPACK13250750)
Packaging Information
spec_VWRC26774.236 spec_VWRC26774.236 spec_VWRC26774.236 spec_VWRC26774.293 spec_VWRC26774.293
Product specification
More
Frequently Bought Together