Order Entry
Denmark
ContactUsLinkComponent
Potassium chromate, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
Potassium chromate, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
Supplier:  Merck
Certificates
Potassium chromate, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
Supplier:  Merck
Certificates

Specification Test Results

  • Solubility:
    637 g/l (20 °C)
  • Melting Point:
    985 °C
  • Molar Mass:
    194.19 g/mol
  • Bulk Density:
    1400 kg/m3
  • Boiling Point:
    1000 °C
  • Density:
    2.73 g/cm3 (18 °C)
  • pH:
    9.0 - 9.8 (50 g/l, H2O, 20 °C)
  • Hazard Class:
    6.1
  • Hazard Class Description:
    Toxic Materials

Specification Test Results

Related Information

Frequently Bought Together