Table clamp for rods
Tempered cast iron, 12/13 mm, Rods
Catalog # BOCH5260
Supplier: BOCHEM
Jump to:
Product Details & Documents
Large table clamp for rods, made of tempered cast iron.
- Can be attached to table tops with a thickness of max. 55 mm
- Green varnished
Specifications
- For:Rods
- Spanning width:12/13 mm
- Material:Tempered cast iron