Order Entry
Denmark
ContactUsLinkComponent
Potassium chloride, Suprapur® Trace_analysis, Supelco®
Potassium chloride, Suprapur® Trace_analysis, Supelco®
Supplier:  Merck
Certificates
Potassium chloride, Suprapur® Trace_analysis, Supelco®
Supplier:  Merck
Certificates

Specifications

  • Environmentally Preferable:

Specifications

Specification Test Results

  • Solubility:
    347 g/l (20 °C)
  • Melting Point:
    773 °C
  • Molar Mass:
    74.55 g/mol
  • Bulk Density:
    1000 kg/m3
  • Boiling Point:
    1413 °C (1013 hPa)
  • Density:
    1.98 g/cm3 (20 °C)
  • pH:
    5.5 - 8.0 (50 g/l, H2O, 25 °C)

Specification Test Results

Related Information

Frequently Bought Together