
Lithium bromide ≥99%, crystalline powder
:7550-35-8
:23303-71-1
:LiBr
:MFCD00011077
:86.84 g/mol
:13,05548
:1265 °C (1013 hPa)
:550 °C
:3.46 g/cm³ (20 °C)
:231-439-8
:GHS07
:Ambient
- Form:White Crystalline Powder
- Storage:Store at room temperature (15 to 30 °C)
Frequently Bought Together