Order Entry
Denmark
ContactUsLinkComponent
Sodium borohydride ≥99%, TraceSELECT™ for the determination of hydride formers by aas, Fluka™
Sodium borohydride ≥99%, TraceSELECT™ for the determination of hydride formers by aas, Fluka™
Certificates
Sodium borohydride ≥99%, TraceSELECT™ for the determination of hydride formers by aas, Fluka™
Certificates

Specification Test Results

  • Assay:
    98.5 - 101.5 %
  • Arsenic (As):
    Max. 0.05 ppm
  • Mercury (Hg):
    Max. 50 ppb
  • Antimony (Sb):
    Max. 0.05 ppm
  • Selenium (Se):
    Max. 0.05 ppm

Specification Test Results

Related Information

Frequently Bought Together