Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Denmark
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Sodium borohydride
Sodium borohydride ≥98%
Sodium borohydride ≥98%
:
Thermo Fisher Scientific
Sodium borohydride ≥98%
:
Thermo Fisher Scientific
:
16940-66-2
:
:
NaBH₄
:
MFCD00003518
:
37.83 g/mol
:
14,8592
:
400 °C (dec.)
:
1.074
:
69 °C
:
Ambient
:
241-004-4
:
1426
:
4.3,I
:
GHS02
:
GHS06
:
GHS05
msds_ALFA088983
Thermo Scientific Acroseal 25 Years
Frequently Bought Together
Acetone ≥99%, TECHNICAL
: VWR Chemicals
2-Propanol, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®
: Merck
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
: VWR Chemicals
Acetonitrile ≥99.9%, LiChrosolv® Reag. Ph. Eur., gradient grade for liquid chromatography, Supelco®
: Merck
Sodium sulphate, anhydrous, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®
: Merck
Methanol ≥99.9% (by GC), LiChrosolv® Reag. Ph. Eur., gradient grade for liquid chromatography, Supelco®
: Merck
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Potassium chloride, EMSURE® for analysis, Supelco®
: Merck
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
: VWR Chemicals
Ethanol absolute ≥99.5%, TechniSolv®, pure
: VWR Chemicals
tri-Sodium citrate dihydrate, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®
: Merck
Potassium sulphate, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®
: Merck
Sodium acetate, anhydrous, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
: Merck
Acetonitrile, anhydrous (max. 0.003% H₂O) ≥99.95%, HiPerSolv CHROMANORM® Reag. USP, Reag. Ph. Eur., ACS, super gradient grade suitable for UPLC/UHPLC instruments
: VWR Chemicals
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Magnesium sulphate heptahydrate, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
: Merck
Sodium chloride 99.5-100.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
: VWR Chemicals
Sodium chloride, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®
: Merck
Sodium borohydride ≥98%
: Thermo Fisher Scientific