Sodium borohydride ≥98%
Leverandør: Thermo Fisher Scientific
Certifikater
Om denne vare
CAS nummer:16940-66-2
:
Formel:NaBH₄
MDL nummer:MFCD00003518
:37.83 g/mol
Merck Index:14,8592
Smeltende Pt:400 °C (dec.)
Tæthed:1.074
Flash Pt:69 °C
Opbevaringstemperatur:Ambient
EINECS:241-004-4
FN:1426
ADR:4.3,I
:GHS02
:GHS06
:GHS05
Relateret information
Frequently Bought Together