Sodium borohydride 99%, powder
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:16940-66-2
:
Formula:NaBH₄
MDL Number:MFCD00003518
:37.83 g/mol
Merck Index:13,08664
Melting Pt:360 °C (dec.)
Density:1,07 g/cm³ (20 °C)
Flash Pt:69 °C
EINECS:241-004-4
UN:1426
ADR:4.3,I
:GHS06
:GHS05
:GHS02
:GHS08
Storage Temperature:Ambient
Specification Test Results
- Appearance:White Powder
- Titration Iodometric:≥98.5 %
Specification Test Results
Frequently Bought Together