
Potassium permanganate 0.02 mol/l (0.1 N) in aqueous solution Reag. Ph. Eur. 3005300
Catalog # CMPL3005300
Supplier: CUSTOM MADE CHEMICALS LAB
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:250 mL
- Seq:2590
Specifications
About this item
:
:158.03 g/mol
Storage Temperature:Ambient
EINECS:231-760-3
UN:3082
ADR:9,III
:GHS09
CAS Number:7722-64-7
Formula:KMnO₄
MDL Number:MFCD00011364