Order Entry
Denmark
ContactUsLinkComponent
Hydrochloric acid 6 M HCl (6N) volumetric solution, Fluka™
Hydrochloric acid 6 M HCl (6N) volumetric solution, Fluka™
Catalog # 72033-1L
CAS Number:  
Hydrochloric acid 6 M HCl (6N) volumetric solution, Fluka™
Catalog # 72033-1L
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Assay:
    5.988 - 6.012 mol/l
  • Calibrated Against:
    tris
  • Vapor Pressure:
    190 mmHg ( 25 °C)

Specification Test Results

About this item

 :
Density:1.1 g/cm³ (20 °C)
EINECS:231-595-7
ADR:8,III
 :36.46 g/mol
UN:1789
 :GHS07
 :GHS05
Storage Temperature:Ambient
CAS Number:7647-01-0
Formula:HCl
MDL Number:MFCD00011324