Order Entry
Denmark
Orders LinkContactUsLinkComponent
 

Tropine

Glass bottle for solids, 10 g
Catalog # ROTH4049.1
Marketsource
Supplier:   Roth Carl
Tropine
Catalog # ROTH4049.1
Supplier:  Roth Carl
Marketsource
CAS Number:  120-29-6
Tropine

Glass bottle for solids, 10 g

CAS Number:  120-29-6
Price
Inquire For Price
Log in for availability
1 * 10 g
Product Details & Documents
Synonyms:  endo-8-Methyl-8-azabicyclo[3.2.1]octan-3-ol
  • CAS Number:120-29-6
  • Formula:C₈H₁₅NO
  • MDL Number:MFCD00180667
  • Molecular Weight:141.21 g/mol
  • EINECS:204-384-2
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    10 g

Specifications