Sodium hydroxide
Glass bottle for solids, 100 g
Catalog # S047
Supplier: G-Biosciences
Jump to:
Product Details & Documents
Synonyms: Caustic soda
- CAS Number:1310-73-2
- Formula:NaOH
- MDL Number:MFCD00003548
- Molecular Weight:40 g/mol
- Merck Index:14,08627
- Boiling Pt:1390 °C (1013 hPa)
- Melting Pt:323 °C
- Density:2.13 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:215-185-5
- UN:1823
- ADR:8,II
- REACH:01-2119457892-27
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g



