Brij® 58 10% in aqueous solution, Proteomics Grade
Supplier: G-Biosciences
Certificates
About this item
CAS Number:9004-95-9
:
Formula:C₅₆H₁₁₄O₂₁
MDL Number:MFCD00080892
:1123.51 g/mol
EINECS:500-014-1
Frequently Bought Together
Pyridine anhydrous, 99.8 % 1 * 2 L
Supplier: Merck
Glycerine ≥98%, GPR RECTAPUR®
Supplier: VWR Chemicals
Adenosine-5’-Triphosphate (ATP) 100 mM in aqueous solution ≥99% for molecular biology
Supplier: Thermo Fisher Scientific
2-Methylbutane 99+%, extra pure
Supplier: Thermo Fisher Scientific
Ethylene glycol 99.5% for analysis
Supplier: Thermo Fisher Scientific
4-(Methylamino)phenol sulphate 99%, extra pure
Supplier: Thermo Fisher Scientific
Toluene (Reag. Ph. Eur.) for analysis, ACS, ISO 1 * 2,5 L
Supplier: PanReac AppliChem
Benzonitrile 99+% for spectroscopy
Supplier: Thermo Fisher Scientific
Petroleum spirit, 60…80 °C
Supplier: Thermo Fisher Scientific
Diethyl ether stabilised, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: Avantor
Diethyl ether ≥99.8% contains inhibitor, Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
Water, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
Tetrabutylammonium dihydrogen phosphate 99% for HPLC
Supplier: Thermo Fisher Scientific
Methanol ≥99.9%, CHROMASOLV™, gradient grade for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals