Diethylene glycol monohexyl ether
Catalog # EHERC12606300
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,25 g
- Seq:3029
Specifications
About this item
:
:190.28 g/mol
EINECS:203-988-3
CAS Number:112-59-4
Formula:C₁₀H₂₂O₃
MDL Number:MFCD00010703