Organic reference standard, 9-Chloroanthracene 50 µg/ml in toluene
Catalog # CAMBULM-8269-1.2
Supplier: CAMBRIDGE ISOTOPE LABS
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1,2 mL
- CAS No.:716-53-0
- Description:9-Chloroanthracene 50 µg/ml in toluene
- Formula:C₁₄H₉Cl
- Description:9-Chloroanthracene